siRNA Details
Gene Symbol: Sh3tc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194344.2 | NP_919325.2 | 7 | 1065 | ||
GenBank mRNA | AK028482.1 | - | 3 | 1161 | ||
AK150064.1 | - | 7 | 843 | |||
AK155330.1 | - | 7 | 826 | |||
AK159089.1 | - | 7 | 844 | |||
AK167283.1 | - | 7 | 766 | |||
AK170383.1 | - | 7 | 898 | |||
AK170396.1 | - | 7 | 845 | |||
AK171209.1 | - | 7 | 822 | |||
AK171242.1 | - | 7 | 822 | |||
BC024909.1 | - | 5 | 515 | |||
BC056461.1 | - | 5 | 409 |
