siRNA Details
Gene Symbol: PDLIM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005451.4 | NP_005442.2 | 13 | 1406 | ||
NM_203352.2 | NP_976227.1 | 13 | 1304 | |||
NR_103804.1 | 13 | 1605 | ||||
XM_011534698.2 | XP_011533000.1 | 12 | 1283 | |||
XM_011534699.2 | XP_011533001.1 | 14 | 1391 | |||
GenBank mRNA | AF265209.1 | - | 13 | 1401 | ||
AF345904.1 | - | 13 | 1362 | |||
AF345905.1 | - | 13 | 1260 | |||
AF345906.1 | - | 14 | 1379 | |||
AK096826.1 | - | 11 | 1996 | |||
BC001093.2 | - | 13 | 1384 | 3506748 | ||
BC067806.1 | - | 13 | 1601 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
