siRNA Details
Gene Symbol: Cep63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081122.1 | NP_001074591.1 | 12 | 1455 | ||
NM_001301689.1 | NP_001288618.1 | 12 | 1455 | |||
GenBank mRNA | BC048718.1 | - | 5 | 607 | ||
KC211185.1 | - | 12 | 1312 |

More Information
Other Species Targeted

Gene Ontology Categories:
