siRNA Details
Gene Symbol: Cep63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081122.1 | NP_001074591.1 | 13 | 1594 | ||
NM_001301689.1 | NP_001288618.1 | 13 | 1594 | |||
GenBank mRNA | AK076549.1 | - | 13 | 1595 | ||
BC048718.1 | - | 6 | 746 | |||
KC211185.1 | - | 13 | 1451 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001244805, NM_001037772, XM_008766491, XM_008766493, XM_008766490, and others.
Gene Ontology Categories:
