siRNA Details
Gene Symbol: Zfyve26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008550.1 | NP_001008550.1 | 11 | 1810 | ||
GenBank mRNA | AK041668.1 | - | 11 | 1807 | ||
AK132344.1 | - | 11 | 1800 | |||
AK220333.1 | - | 11 | 1781 | |||
BC141359.1 | - | 11 | 1793 | 9055971 |

More Information
Other Species Targeted

Gene Ontology Categories:
