siRNA Details
Gene Symbol: Zfyve26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008550.1 | NP_001008550.1 | 11 | 2120 | ||
GenBank mRNA | AK041668.1 | - | 11 | 2117 | ||
AK132344.1 | - | 11 | 2110 | |||
AK220333.1 | - | 11 | 2091 | |||
BC141359.1 | - | 11 | 2103 | 9055971 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
