siRNA Details
Gene Symbol: 4921509C19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198655.2 | NP_941057.1 | 2 | 301 | ||
GenBank mRNA | AK029504.1 | - | 2 | 303 | ||
BC119071.1 | - | 1 | 131 | 8733820 | ||
BC119073.1 | - | 1 | 131 | 8733822 |
