siRNA Details
Gene Symbol: Oog2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198661.3 | NP_941063.1 | 4 | 486 | ||
GenBank mRNA | AK139660.1 | - | 2 | 1481 | ||
AY573562.1 | - | 4 | 486 | |||
BC052839.1 | - | 4 | 486 | 30050073 |
Gene Symbol: Gm13084 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005371.3 | NP_001005371.2 | 3 | 424 | ||
GenBank mRNA | AK145369.1 | - | 3 | 421 | ||
BC080785.1 | - | 3 | 422 | 30481291 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
