siRNA Details
Gene Symbol: Tbc1d31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081396.2 | NP_001074865.2 | 6 | 895 | ||
NM_001167679.1 | NP_001161151.1 | 6 | 895 | |||
GenBank mRNA | AB185844.1 | - | 6 | 863 | ||
AK084542.1 | - | 5 | 659 | |||
AK141805.1 | - | 6 | 765 | |||
AK154264.1 | - | 6 | 860 | |||
AK161403.1 | - | 5 | 712 | |||
AK171024.1 | - | 6 | 860 | |||
BC050039.1 | - | 6 | 841 | 30016266 | ||
BC066830.1 | - | 6 | 827 | 30463017 |

More Information
Other Species Targeted

Gene Ontology Categories:
