siRNA Details
Gene Symbol: Zfp652 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201609.2 | NP_963903.2 | 1 | 705 | ||
GenBank mRNA | AK220361.1 | - | 1 | 550 | ||
BC055752.1 | - | 1 | 705 | 6414140 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
