siRNA Details
Gene Symbol: Adora1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008533.3 | NP_001008533.1 | 2 | 3241 | ||
NM_001039510.2 | NP_001034599.1 | 3 | 2426 | |||
NM_001282945.1 | NP_001269874.1 | 3 | 2460 | |||
NM_001291928.1 | NP_001278857.1 | 2 | 1992 | |||
NM_001291930.1 | NP_001278859.1 | 2 | 2028 | |||
GenBank mRNA | AK043954.1 | - | 2 | 3183 | ||
AK045729.1 | - | 2 | 2781 | |||
AK134532.1 | - | 3 | 2435 | |||
AK134709.1 | - | 2 | 2607 | |||
BC026602.1 | - | 2 | 2102 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
