siRNA Details
Gene Symbol: Nfatc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010901.2 | NP_035031.2 | 3 | 1632 | ||
GenBank mRNA | AK039929.1 | - | 3 | 1549 | ||
AK051602.1 | - | 3 | 1590 | |||
AK131919.1 | - | 1 | 44 | |||
AK134054.1 | - | 3 | 1551 | |||
AK147708.1 | - | 3 | 1603 | |||
AK155087.1 | - | 3 | 1602 | |||
AK220187.1 | - | 3 | 1404 | |||
BC120900.1 | - | 3 | 1351 | 40085687 | ||
BC120901.1 | - | 3 | 1351 | 40085688 | ||
D85612.1 | - | 3 | 1348 | |||
EU887626.1 | - | 3 | 1320 | |||
EU887627.1 | - | 3 | 1320 | |||
EU887628.1 | - | 3 | 1320 | |||
EU887629.1 | - | 3 | 1344 | |||
EU887630.1 | - | 3 | 1344 | |||
EU887631.1 | - | 3 | 1344 | |||
U28807.1 | - | 3 | 1311 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001108447, XM_008772519, XM_008772520, XM_008772518, XM_006255513.
Gene Ontology Categories:
