siRNA Details
Gene Symbol: Nfatc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010901.2 | NP_035031.2 | 3 | 1614 | ||
GenBank mRNA | AK039929.1 | - | 3 | 1531 | ||
AK051602.1 | - | 3 | 1572 | |||
AK131919.1 | - | 1 | 26 | |||
AK134054.1 | - | 3 | 1533 | |||
AK147708.1 | - | 3 | 1585 | |||
AK155087.1 | - | 3 | 1584 | |||
AK220187.1 | - | 3 | 1386 | |||
BC120900.1 | - | 3 | 1333 | 40085687 | ||
BC120901.1 | - | 3 | 1333 | 40085688 | ||
D85612.1 | - | 3 | 1330 | |||
EU887626.1 | - | 3 | 1302 | |||
EU887627.1 | - | 3 | 1302 | |||
EU887628.1 | - | 3 | 1302 | |||
EU887629.1 | - | 3 | 1326 | |||
EU887630.1 | - | 3 | 1326 | |||
EU887631.1 | - | 3 | 1326 | |||
U28807.1 | - | 3 | 1293 |

More Information
Other Species Targeted

Gene Ontology Categories:
