siRNA Details
Gene Symbol: MAP3K6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297609.1 | NP_001284538.1 | 6 | 1336 | ||
NM_004672.4 | NP_004663.3 | 7 | 1360 | |||
XM_017002771.1 | XP_016858260.1 | 5 | 729 | |||
XM_017002772.1 | XP_016858261.1 | 7 | 2531 | |||
GenBank mRNA | AB167411.1 | - | 7 | 1091 | ||
AB208805.1 | - | 6 | 805 | |||
AF100318.1 | - | 4 | 505 | |||
BC015914.1 | - | 3 | 262 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
