siRNA Details
Gene Symbol: Soga3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026138.2 | NP_080414.2 | 3 | 1716 | ||
GenBank mRNA | AK018139.1 | - | 2 | 246 | ||
AK030682.1 | - | 3 | 1589 | |||
AK044134.1 | - | 2 | 571 | |||
AK163577.1 | - | 2 | 216 | |||
BC066079.1 | - | 3 | 1873 | 30355520 |
