siRNA Details
Gene Symbol: Ppih | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110129.1 | NP_001103599.1 | 3, 4 | 200 | ||
NM_001110130.1 | NP_001103600.1 | 3, 4 | 200 | |||
NM_028677.4 | NP_082953.1 | 3, 4 | 200 | |||
GenBank mRNA | AK003179.1 | - | 3, 4 | 178 | ||
AK005202.1 | - | 3, 4 | 161 | |||
AK008394.1 | - | 3, 4 | 178 | |||
AK014665.1 | - | 3, 4 | 161 | |||
AK134995.1 | - | 3, 4 | 180 | |||
AK136330.1 | - | 3, 4 | 159 | |||
AK164299.1 | - | 3, 4 | 165 | |||
BC050116.1 | - | 3, 4 | 154 | 6432881 | ||
BY708305.1 | - | 3, 4 | 177 | |||
CJ071503.1 | - | 3, 4 | 223 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
