siRNA Details
Gene Symbol: Ppih | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028677.4 | NP_082953.1 | 10 | 732 | ||
GenBank mRNA | AK003179.1 | - | 10 | 710 | ||
AK005202.1 | - | 10 | 712 | |||
BC050116.1 | - | 10 | 686 | 6432881 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
