siRNA Details
Gene Symbol: MAP3K13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242314.1 | NP_001229243.1 | 3 | 696 | ||
NM_004721.4 | NP_004712.1 | 2 | 811 | |||
XM_011513310.2 | XP_011511612.1 | 2 | 969 | |||
XM_017007456.1 | XP_016862945.1 | 3 | 707 | |||
GenBank mRNA | AB001872.1 | - | 2 | 677 | ||
AK296961.1 | - | 2 | 739 | |||
AK312714.1 | - | 3 | 661 | |||
BC111726.1 | - | 2 | 617 | 40028559 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
