siRNA Details
Gene Symbol: Phlpp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133821.3 | NP_598582.3 | 3, 4 | 1910 | ||
GenBank mRNA | AB093251.1 | - | 3, 4 | 669 | ||
BC059254.1 | - | 3, 4 | 570 | 6406203 | ||
CN525246.1 | - | 3, 4 | 376 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
