siRNA Details
Gene Symbol: Taok1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144825.2 | NP_659074.2 | 16 | 2542 | ||
GenBank mRNA | BC016522.1 | - | 4 | 536 | ||
BC047271.1 | - | 15 | 1889 | |||
BC151012.1 | - | 16 | 2336 | 9087925 | ||
BC151015.1 | - | 16 | 2336 | 9087928 | ||
BC151016.1 | - | 16 | 2335 | 9087929 | ||
CN717576.1 | - | 3 | 350 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
