siRNA Details
Gene Symbol: Taok1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144825.2 | NP_659074.2 | 20 | 3811 | ||
GenBank mRNA | AK173156.1 | - | 4 | 1145 | ||
BC016522.1 | - | 8 | 1805 | |||
BC034906.1 | - | 2 | 654 | |||
BC047271.1 | - | 19 | 3158 |

More Information
Other Species Targeted

Gene Ontology Categories:
