siRNA Details
Gene Symbol: Olfr657 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146312.2 | NP_666424.2 | 1 | 138 | ||
GenBank mRNA | BC104311.1 | - | 1 | 133 |
Gene Symbol: Olfr504 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011858.1 | NP_001011858.1 | 1 | 138 | ||
GenBank mRNA | BC104310.1 | - | 1 | 138 | 40044370 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
