siRNA Details
Gene Symbol: Olfr958 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146330.1 | NP_666442.1 | 1 | 604 | ||
GenBank mRNA | BC148230.1 | - | 1 | 608 | 40142225 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olfr958 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146330.1 | NP_666442.1 | 1 | 604 | ||
GenBank mRNA | BC148230.1 | - | 1 | 608 | 40142225 |