siRNA Details
Gene Symbol: KRT7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF509887.1 | - | 5 | 859 | ||
AK025881.1 | - | 5 | 857 | |||
AK128505.1 | - | 5 | 749 | |||
AK128848.1 | - | 6 | 785 | |||
AK129924.1 | - | 6 | 864 | |||
BC002700.2 | - | 5 | 845 | 3610347 | ||
BC009521.1 | - | 1 | 17 | |||
X03212.1 | - | 5 | 864 | |||
Z36852.1 | - | 3 | 270 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
