siRNA Details
Gene Symbol: Olfr1512 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146432.2 | NP_666643.1 | 1 | 489 | ||
GenBank mRNA | BC133656.1 | - | 1 | 489 | 40042187 |
Gene Symbol: Olfr1513 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012269.2 | NP_001012269.2 | 1 | 489 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
