siRNA Details
Gene Symbol: Olfr203 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146486.2 | NP_666697.2 | 1 | 578 | ||
GenBank mRNA | BC128025.1 | - | 1 | 581 | 40136560 |
Gene Symbol: Olfr194 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005524.2 | NP_001005524.2 | 1 | 578 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
