siRNA Details
Gene Symbol: Olfr419 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146715.2 | NP_666926.2 | 1 | 849 | ||
GenBank mRNA | BC119050.1 | - | 1 | 849 | 8733799 | |
BC119052.1 | - | 1 | 849 | 8733801 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001000911.
Gene Ontology Categories:
