siRNA Details
Gene Symbol: LIMK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK093554.1 | - | 5 | 591 | ||
AK293774.1 | - | 10 | 1414 | |||
AK299112.1 | - | 9 | 1516 | |||
AK302553.1 | - | 10 | 1250 | |||
AK314391.1 | - | 9 | 1469 | |||
AL117466.1 | - | 9 | 1451 | |||
CR456513.1 | - | 10 | 1232 | |||
D45906.1 | - | 10 | 1313 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
