siRNA Details
Gene Symbol: NACA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199290.3 | NP_954984.1 | 1 | 264 | ||
GenBank mRNA | BC062710.1 | - | 1 | 268 | 6614481 |

More Information
Other Species Targeted

Gene Ontology Categories:
