siRNA Details
Gene Symbol: Olfr658 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147049.4 | NP_667260.1 | 2 | 1377 | ||
GenBank mRNA | AK033992.1 | - | 2 | 1378 | ||
BC104120.2 | - | 1 | 956 | 40044139 | ||
BC104121.1 | - | 1 | 956 |
Gene Symbol: Olfr503 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011527.1 | NP_001011527.1 | 1 | 956 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
