siRNA Details
Gene Symbol: Phldb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252442.1 | NP_001239371.1 | 2 | 1057 | ||
NM_153412.4 | NP_700461.2 | 2 | 1057 | |||
GenBank mRNA | AF506821.1 | - | 2 | 1058 | ||
AK049595.1 | - | 2 | 911 | |||
AK050493.1 | - | 3 | 1224 | |||
AK165251.1 | - | 2 | 1019 | |||
BC050915.1 | - | 2 | 913 | 6405379 | ||
BC060683.1 | - | 2 | 913 | 6405379 |
