siRNA Details
Gene Symbol: Bclaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025392.1 | NP_001020563.1 | 10, 11 | 2644 | ||
NM_153787.2 | NP_722482.1 | 10, 11 | 2638 | |||
GenBank mRNA | AK079110.1 | - | 6, 7 | 894 | ||
AK129071.1 | - | 7, 8 | 1928 | |||
AK149724.1 | - | 10, 11 | 2597 | |||
AK150017.1 | - | 10, 11 | 2597 | |||
BC029682.1 | - | 10, 11 | 2576 | |||
BC032973.1 | - | 1, 2 | 319 | |||
BC034300.1 | - | 10, 11 | 2570 | 4507876 | ||
CN530237.1 | - | 1, 2 | 29 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
