siRNA Details
Gene Symbol: RTN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005619.4 | NP_005610.1 | 2, 3 | 320 | ||
NM_206900.2 | NP_996783.1 | 2, 3 | 320 | |||
GenBank mRNA | AF004222.1 | - | 2, 3 | 219 | ||
AK314158.1 | - | 2, 3 | 303 | |||
BM548456.1 | - | 315 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
