siRNA Details
Gene Symbol: Abat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170978.1 | NP_001164449.1 | 3 | 329 | ||
NM_172961.3 | NP_766549.2 | 3 | 329 | |||
GenBank mRNA | AK036128.1 | - | 3 | 274 | ||
AK083062.1 | - | 3 | 276 | |||
AK132282.1 | - | 3 | 278 | |||
AK160802.1 | - | 3 | 277 | |||
AK165348.1 | - | 3 | 279 | |||
BC058079.1 | - | 3 | 269 | 6826741 | ||
BC058521.1 | - | 3 | 269 | 6825487 | ||
CX237932.1 | - | 3 | 329 |
