siRNA Details
Gene Symbol: Abat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170978.1 | NP_001164449.1 | 15 | 1583 | ||
NM_172961.3 | NP_766549.2 | 16 | 1751 | |||
GenBank mRNA | AK036128.1 | - | 15 | 1528 | ||
AK132282.1 | - | 16 | 1700 | |||
AK156736.1 | - | 4 | 460 | |||
AK165348.1 | - | 16 | 1702 | |||
BC058079.1 | - | 16 | 1691 | 6826741 | ||
BC058521.1 | - | 16 | 1691 | 6825487 |
