siRNA Details
Gene Symbol: SLC6A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142805.1 | NP_001136277.1 | 1, 2 | 548 | ||
NM_005629.3 | NP_005620.1 | 1, 2 | 548 | |||
GenBank mRNA | AB209704.1 | - | 1, 2 | 352 | ||
BC012355.1 | - | 1, 2 | 315 | 4634609 | ||
BC081558.1 | - | 1, 2 | 549 | 4823748 | ||
KJ534942.1 | - | 1, 2 | 270 | |||
KJ534943.1 | - | 1, 2 | 270 | |||
KJ534944.1 | - | 1, 2 | 270 | |||
KJ534945.1 | - | 1, 2 | 270 | |||
L31409.1 | - | 1, 2 | 743 | |||
S74039.1 | - | 1, 2 | 288 |
Gene Symbol: SLC6A10P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003083.2 | 1, 2 | 241 | |||
GenBank mRNA | AK125843.1 | - | 1, 2 | 242 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
