siRNA Details
Gene Symbol: SSX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005636.3 | NP_005627.1 | 8 | 1003 | ||
NM_175729.1 | NP_783856.1 | 7 | 867 | |||
XM_017029759.1 | XP_016885248.1 | 8 | 1061 | |||
GenBank mRNA | BC005325.1 | - | 8 | 1003 | 3961688 |
Gene Symbol: SSX4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034832.3 | NP_001030004.1 | 8 | 1014 | ||
NM_001040612.2 | NP_001035702.1 | 7 | 878 | |||
XM_017029613.1 | XP_016885102.1 | 8 | 1061 |
Gene Symbol: SSX6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028366.1 | 8 | 1073 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
