siRNA Details
Gene Symbol: DYRK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003583.3 | NP_003574.1 | 2 | 2245 | ||
NM_006482.2 | NP_006473.2 | 3 | 2394 | |||
XM_017020032.1 | XP_016875521.1 | 2 | 2537 | |||
GenBank mRNA | BC005809.2 | - | 2 | 2199 | 3831977 | |
DA768075.1 | - | 1 | 24 | |||
Y13493.1 | - | 3 | 2152 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
