siRNA Details
Gene Symbol: SLC35A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042498.2 | NP_001035963.1 | 3, 4 | 756 | ||
NM_001282649.1 | NP_001269578.1 | 2, 3 | 573 | |||
NM_001282650.1 | NP_001269579.1 | 3, 4 | 795 | |||
NM_001282651.1 | NP_001269580.1 | 4, 5 | 840 | |||
NM_005660.2 | NP_005651.1 | 3, 4 | 756 | |||
GenBank mRNA | AK225508.1 | - | 3, 4 | 444 | ||
AK292816.1 | - | 3, 4 | 473 | |||
AK293415.1 | - | 2, 3 | 261 | |||
AK293419.1 | - | 3, 4 | 483 | |||
AK298484.1 | - | 4, 5 | 528 | |||
CR450287.1 | - | 3, 4 | 434 | |||
D84454.1 | - | 3, 4 | 757 | |||
D88146.1 | - | 3, 4 | 438 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
