siRNA Details
Gene Symbol: Chd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201637.2 | NP_963999.2 | 21 | 4209 | ||
GenBank mRNA | AK141273.1 | - | 4 | 350 | ||
BC049854.1 | - | 3 | 165 | |||
BC057382.1 | - | 3 | 298 | |||
DQ190419.1 | - | 21 | 4209 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
