siRNA Details
Gene Symbol: Chd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201637.2 | NP_963999.2 | 22 | 4427 | ||
GenBank mRNA | AK141273.1 | - | 5 | 568 | ||
BC049854.1 | - | 4 | 383 | |||
BC057382.1 | - | 4 | 516 | |||
DQ190419.1 | - | 22 | 4427 |

More Information
Other Species Targeted

Gene Ontology Categories:
