siRNA Details
Gene Symbol: Chd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201637.2 | NP_963999.2 | 34 | 6815 | ||
GenBank mRNA | AK031711.1 | - | 4 | 1917 | ||
AK080266.1 | - | 3 | 536 | |||
AK129393.1 | - | 4 | 2043 | |||
AK141273.1 | - | 17 | 2956 | |||
AK160299.1 | - | 4 | 758 | |||
BC023856.1 | - | 1 | 259 | |||
BC049854.1 | - | 16 | 2795 | |||
BC057382.1 | - | 16 | 2904 | |||
DQ190419.1 | - | 34 | 6815 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
