siRNA Details
Gene Symbol: Pld1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164056.1 | NP_001157528.1 | 3, 4 | 420 | ||
NM_008875.4 | NP_032901.2 | 2, 3 | 365 | |||
GenBank mRNA | BC068144.1 | - | 2, 3 | 222 | 30532470 | |
CF727963.1 | - | 2, 3 | 365 | |||
U87868.1 | - | 3, 4 | 403 |
