siRNA Details
Gene Symbol: Edil3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037987.3 | NP_001033076.1 | 11 | 3519 | ||
NM_010103.4 | NP_034233.1 | 10 | 3489 | |||
GenBank mRNA | AK081003.1 | - | 10 | 3322 | ||
AK139093.1 | - | 11 | 3481 | |||
AK141907.1 | - | 11 | 3502 | |||
BC056386.1 | - | 11 | 3419 | 6854497 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
