siRNA Details
Gene Symbol: UBXN8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282189.1 | NP_001269118.1 | 3, 4 | 204 | ||
NM_005671.3 | NP_005662.1 | 3, 4 | 315 | |||
XM_011544655.2 | XP_011542957.1 | 5, 6 | 1392 | |||
XM_017013862.1 | XP_016869351.1 | 4, 5 | 504 | |||
GenBank mRNA | AK296600.1 | - | 3, 4 | 489 | ||
AY302140.1 | - | 3, 4 | 179 | |||
BC020694.1 | - | 3, 4 | 299 | 4772692 | ||
BI462115.1 | - | 3, 4 | 320 | |||
D83767.1 | - | 3, 4 | 305 |
