siRNA Details
Gene Symbol: Zfp106 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011743.2 | NP_035873.2 | 5 | 658 | ||
GenBank mRNA | AK089359.1 | - | 5 | 656 | ||
AK147887.1 | - | 5 | 659 | |||
AK155019.1 | - | 5 | 656 | |||
BC010440.1 | - | 3 | 300 | |||
BC025424.1 | - | 5 | 647 | 4459713 | ||
U59739.1 | - | 2 | 725 |
