siRNA Details
Gene Symbol: Chka | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271496.1 | NP_001258425.1 | 12 | 1767 | ||
NM_013490.4 | NP_038518.2 | 11 | 1713 | |||
GenBank mRNA | AB011002.1 | - | 11 | 1572 | ||
AK166465.1 | - | 11 | 1654 | |||
BC004719.1 | - | 9 | 903 | |||
BC060218.1 | - | 10 | 3793 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
