siRNA Details
Gene Symbol: Chka | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271496.1 | NP_001258425.1 | 12 | 2484 | ||
NM_013490.4 | NP_038518.2 | 11 | 2430 | |||
GenBank mRNA | AB011002.1 | - | 11 | 2289 | ||
AK005057.1 | - | 13 | 2545 | |||
BC060218.1 | - | 10 | 4510 | |||
BU918359.1 | - | 1 | 100 |

More Information
Other Species Targeted

Gene Ontology Categories:
