siRNA Details
Gene Symbol: Chka | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271496.1 | NP_001258425.1 | 12 | 1875 | ||
NM_013490.4 | NP_038518.2 | 11 | 1821 | |||
GenBank mRNA | AB011002.1 | - | 11 | 1680 | ||
BC060218.1 | - | 10 | 3901 |

More Information
Other Species Targeted

Gene Ontology Categories:
