siRNA Details
Gene Symbol: Cmya5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023821.3 | NP_076310.2 | 2 | 1047 | ||
GenBank mRNA | AJ511265.1 | - | 1 | 724 | ||
AJ575748.1 | - | 1047 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
